Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145931.2 | NP_666043.2 | 10 | 1254 | ||
GenBank mRNA | AK028438.1 | - | 10 | 1179 | ||
AK040507.1 | - | 10 | 1202 | |||
AK049834.1 | - | 10 | 1189 | |||
AK052830.1 | - | 11 | 1365 | |||
AK150952.1 | - | 10 | 1173 | |||
AK166876.1 | - | 10 | 1202 | |||
AK167758.1 | - | 10 | 1203 | |||
BC027330.1 | - | 10 | 1150 | 3674968 | ||
BC029320.1 | - | 10 | 1152 |