Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145932.3 | NP_666044.1 | 1 | 370 | ||
GenBank mRNA | AK052725.1 | - | 1 | 371 | ||
AK147155.1 | - | 190 | ||||
BC024441.1 | - | 1 | 178 | 4973857 | ||
BC025912.1 | - | 1 | 181 | 5064489 | ||
BC031178.1 | - | 1 | 121 | 4988213 |