Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026494.3 | NP_080770.2 | 2 | 650 | ||
GenBank mRNA | AK006290.1 | - | 2 | 162 | ||
AK018256.1 | - | 2 | 622 | |||
AK159952.1 | - | 2 | 650 | |||
AK182947.1 | - | 1 | 48 | |||
BC021894.1 | - | 2 | 587 | 3494382 |