Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026494.3 | NP_080770.2 | 3 | 938 | ||
GenBank mRNA | AK006290.1 | - | 3 | 450 | ||
AK018256.1 | - | 3 | 910 | |||
AK159952.1 | - | 3 | 938 | |||
AK213637.1 | - | 1 | 165 | |||
BC021894.1 | - | 3 | 875 | 3494382 |