Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134122.2 | NP_598883.1 | 3, 4 | 572 | ||
GenBank mRNA | BC021750.1 | - | 3, 4 | 575 | 4503599 | |
BC026576.1 | - | 3, 4 | 544 | 4984796 |