Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174989.4 | NP_778154.1 | 2 | 627 | ||
GenBank mRNA | AB091053.1 | - | 1 | 494 | ||
AK054219.1 | - | 2 | 587 | |||
AK083836.1 | - | 2 | 605 | |||
AK086237.1 | - | 2 | 605 | |||
AK089679.1 | - | 2 | 605 | |||
AK143766.1 | - | 2 | 619 | |||
AK150150.1 | - | 2 | 621 | |||
AK155245.1 | - | 2 | 627 | |||
AK155906.1 | - | 2 | 619 | |||
AK171578.1 | - | 2 | 619 | |||
BC037048.1 | - | 2 | 627 | |||
BC094338.1 | - | 1 | 563 | 30606722 |