Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289490.1 | NP_001276419.1 | 4 | 633 | ||
NM_001289491.1 | NP_001276420.1 | 4 | 358 | |||
NM_207219.4 | NP_997102.2 | 4 | 498 | |||
GenBank mRNA | AK148136.1 | - | 4 | 620 | ||
AK153512.1 | - | 4 | 421 | |||
BC022574.1 | - | 3 | 277 | 3989373 | ||
BC157987.1 | - | 4 | 383 | 9007303 | ||
BY741306.1 | - | 4 | 620 |