Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134134.2 | NP_598895.2 | 20 | 3664 | ||
NM_178277.1 | NP_840061.1 | 19 | 3759 | |||
GenBank mRNA | AB045322.1 | - | 20 | 3665 | ||
AK129085.1 | - | 16 | 2667 | |||
AK132529.1 | - | 13 | 2143 | |||
AK144993.1 | - | 17 | 3175 | |||
BC025148.1 | - | 5 | 627 | |||
BC058089.1 | - | 17 | 3057 | |||
BC060046.1 | - | 13 | 2089 | |||
BC066029.1 | - | 14 | 2895 | 6417061 | ||
BC079580.1 | - | 19 | 3707 | 5702202 |