Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgxb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134134.2 | NP_598895.2 | 21 | 4032 | ||
NM_178277.1 | NP_840061.1 | 20 | 4127 | |||
GenBank mRNA | AB045322.1 | - | 21 | 4033 | ||
AK129085.1 | - | 17 | 3035 | |||
AK132529.1 | - | 14 | 2511 | |||
AK144993.1 | - | 18 | 3543 | |||
BC025148.1 | - | 6 | 995 | |||
BC038134.1 | - | 1 | 123 | |||
BC058089.1 | - | 18 | 3425 | |||
BC060046.1 | - | 14 | 2457 | |||
BC066029.1 | - | 15 | 3263 | 6417061 | ||
BC079580.1 | - | 20 | 4075 | 5702202 |