Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205336.1 | NP_001192265.1 | 14 | 2280 | ||
NM_139206.2 | NP_631945.2 | 14 | 2080 | |||
GenBank mRNA | AF469621.1 | - | 13 | 2030 | ||
AF469622.1 | - | 14 | 2221 | |||
AK036638.1 | - | 14 | 2014 | |||
AK053122.1 | - | 13 | 2470 | |||
AK220486.1 | - | 13 | 2509 | |||
BC068145.1 | - | 14 | 2068 | 6856630 |