Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139143.3 | NP_631882.2 | 9 | 2664 | ||
GenBank mRNA | AB071697.1 | - | 8 | 2047 | ||
AK028976.1 | - | 9 | 2623 | |||
BC054780.1 | - | 8 | 1577 | 6813896 | ||
BC055012.1 | - | 9 | 2334 | 6337369 |