Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134139.1 | NP_598900.1 | 3 | 233 | ||
GenBank mRNA | AK077521.1 | - | 3 | 234 | ||
AK145704.1 | - | 3 | 246 | |||
BC019968.1 | - | 3 | 233 | 4167260 |