Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080706.1 | NP_001074175.1 | 20 | 2927 | ||
GenBank mRNA | AB287295.1 | - | 20 | 2652 | ||
AK141888.1 | - | 14 | 3374 | |||
BC094345.1 | - | 1 | 44 |