Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178930.3 | NP_849261.2 | 32 | 4408 | ||
GenBank mRNA | AK172919.1 | - | 29 | 4001 | ||
BC023791.1 | - | 6 | 826 | |||
BC024129.1 | - | 15 | 2066 | |||
BC026621.1 | - | 3 | 440 | |||
BC034745.1 | - | 17 | 2173 | |||
BC059883.1 | - | 9 | 1073 | |||
BC060728.1 | - | 24 | 3828 | |||
BC076569.1 | - | 32 | 4408 | 6409170 | ||
BC082336.1 | - | 33 | 4403 |