Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175353.2 | NP_780562.2 | 16 | 1632 | ||
GenBank mRNA | AK082881.1 | - | 16 | 1611 | ||
AK151524.1 | - | 16 | 1633 | |||
AK151838.1 | - | 16 | 1633 | |||
BC025616.1 | - | 3 | 206 | |||
BC026859.1 | - | 16 | 1585 | 2631201 |