Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam111a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026640.2 | NP_080916.1 | 4 | 961 | ||
GenBank mRNA | AK019499.1 | - | 4 | 962 | ||
AK045189.1 | - | 4 | 963 | |||
AK142496.1 | - | 3 | 761 | |||
AK172402.1 | - | 4 | 974 | |||
BC038020.1 | - | 4 | 955 | 3599082 |