Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029735.1 | NP_084011.1 | 10 | 1265 | ||
GenBank mRNA | AK028687.1 | - | 3 | 286 | ||
AK141072.1 | - | 10 | 1320 | |||
AK148463.1 | - | 10 | 1226 | |||
AK160986.1 | - | 10 | 1286 | |||
BC040802.1 | - | 10 | 1263 | |||
BC058337.1 | - | 10 | 1218 | |||
BC094679.1 | - | 9 | 1059 | |||
BC141049.1 | - | 10 | 1184 | 9055661 |