Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276292.1 | NP_001263221.1 | 8 | 1064 | ||
NM_177327.5 | NP_796301.2 | 9 | 1226 | |||
GenBank mRNA | AK033138.1 | - | 1 | 40 | ||
AK137588.1 | - | 9 | 1151 | |||
AK168567.1 | - | 9 | 1177 | |||
BC026829.1 | - | 8 | 1002 | 4209397 | ||
BC150812.1 | - | 8 | 1061 | 9087723 |