Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036243.1 | - | 3 | 559 | ||
AK047566.1 | - | 3 | 624 | |||
AK049519.1 | - | 3 | 613 | |||
AK081690.1 | - | 3 | 584 | |||
AK147139.1 | - | 3 | 559 | |||
AK163314.1 | - | 3 | 458 | |||
AK171925.1 | - | 3 | 456 | |||
AK184304.1 | - | 2 | 153 | |||
AK207307.1 | - | 2 | 153 | |||
AK217012.1 | - | 2 | 153 | |||
AK218390.1 | - | 2 | 344 | |||
BC016406.1 | - | 3 | 546 | 4489112 | ||
BC017547.1 | - | 3 | 435 | 4920702 |