Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130860.3 | NP_570930.1 | 6 | 797 | ||
GenBank mRNA | AF327431.1 | - | 6 | 669 | ||
AK089276.1 | - | 6 | 636 | |||
AK142397.1 | - | 4 | 832 | |||
AK143217.1 | - | 6 | 798 | |||
AK144607.1 | - | 6 | 778 | |||
AK155733.1 | - | 4 | 1165 | |||
AK157340.1 | - | 6 | 769 | |||
AK187471.1 | - | 1 | 60 | |||
BC003901.1 | - | 6 | 742 | 3601310 |