Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130860.3 | NP_570930.1 | 5 | 662 | ||
GenBank mRNA | AF327431.1 | - | 5 | 534 | ||
AK089276.1 | - | 5 | 501 | |||
AK142397.1 | - | 3 | 697 | |||
AK143217.1 | - | 5 | 663 | |||
AK144607.1 | - | 5 | 643 | |||
AK155733.1 | - | 4 | 873 | |||
AK157340.1 | - | 5 | 634 | |||
BC003901.1 | - | 5 | 607 | 3601310 |