Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081363.2 | NP_001074832.2 | 13 | 5769 | ||
GenBank mRNA | AF194970.1 | - | 2 | 4054 | ||
AK157703.1 | - | 2 | 885 | |||
AK163859.1 | - | 1 | 323 | |||
DQ642022.1 | - | 12 | 5626 |