Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111277.1 | NP_001104747.1 | 7 | 761 | ||
NM_175135.4 | NP_780344.2 | 7 | 761 | |||
GenBank mRNA | AK145856.1 | - | 7 | 738 | ||
AK172173.1 | - | 6 | 643 | |||
AK202868.1 | - | 2 | 53 | |||
BC115657.1 | - | 5 | 435 | 40103205 | ||
BC115658.1 | - | 5 | 435 | 40103206 |