Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113549.1 | NP_001107021.1 | 28 | 4521 | ||
NM_175641.2 | NP_783572.2 | 31 | 4720 | |||
GenBank mRNA | AF410798.1 | - | 31 | 4801 | ||
AF410799.1 | - | 28 | 4497 | |||
AK009837.1 | - | 2 | 502 | |||
AK054297.1 | - | 28 | 4488 | |||
AK157031.1 | - | 28 | 4537 | |||
BC022658.1 | - | 2 | 433 | |||
BC032073.1 | - | 29 | 4473 | |||
BC059016.1 | - | 28 | 4498 | |||
BC141005.1 | - | 31 | 4726 | 9055617 |