Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054051.1 | NP_473392.1 | 6 | 777 | ||
GenBank mRNA | AB054987.1 | - | 2 | 170 | ||
AK089414.1 | - | 6 | 767 | |||
AK146559.1 | - | 6 | 827 | |||
AK170392.1 | - | 6 | 704 | |||
AK171497.1 | - | 6 | 767 | |||
AY050219.1 | - | 6 | 606 | |||
BC047282.1 | - | 6 | 777 | 6491155 |