Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181405.4 | NP_852070.3 | 7 | 1474 | ||
GenBank mRNA | AK075626.1 | - | 3 | 805 | ||
AK154881.1 | - | 5 | 2048 | |||
BC031789.1 | - | 7 | 894 | 4006916 | ||
CX218581.1 | - | 3 | 360 |