Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181405.4 | NP_852070.3 | 11 | 2078 | ||
GenBank mRNA | AK075626.1 | - | 7 | 1344 | ||
AK087430.1 | - | 5 | 581 | |||
AK138067.1 | - | 11 | ||||
AK154881.1 | - | 8 | 3398 | |||
BC024079.1 | - | 5 | 591 | |||
BC031789.1 | - | 11 | 1498 | 4006916 | ||
CA750931.1 | - | 3 | 445 |