Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175358.4 | NP_780567.1 | 5 | 664 | ||
GenBank mRNA | AK029137.1 | - | 5 | 404 | ||
AK032922.1 | - | 5 | 385 | |||
AK077949.1 | - | 5 | 413 | |||
AK132304.1 | - | 5 | 665 | |||
AK160360.1 | - | 5 | 404 |