Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133805.3 | NP_598566.3 | 5 | 485 | ||
GenBank mRNA | AF482000.1 | - | 5 | 946 | ||
AK079111.1 | - | 5 | 419 | |||
AK079522.1 | - | 5 | 433 | |||
AK088323.1 | - | 5 | 405 | |||
AK090013.1 | - | 5 | 414 | |||
AK142989.1 | - | 4 | 1454 | |||
AK145899.1 | - | 5 | 412 | |||
AK162502.1 | - | 5 | 420 | |||
AK195692.1 | - | 4 | 260 | |||
BC017690.1 | - | 5 | 453 | 4238947 | ||
BC021488.1 | - | 5 | 444 | 5344864 | ||
BC024421.1 | - | 5 | 438 | 4987584 |