Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133805.3 | NP_598566.3 | 3, 4 | 316 | ||
GenBank mRNA | AF482000.1 | - | 3, 4 | 777 | ||
AK079111.1 | - | 3, 4 | 250 | |||
AK079522.1 | - | 3, 4 | 264 | |||
AK088323.1 | - | 3, 4 | 236 | |||
AK090013.1 | - | 3, 4 | 245 | |||
AK142989.1 | - | 2, 3 | 1285 | |||
AK145899.1 | - | 3, 4 | 243 | |||
AK162502.1 | - | 3, 4 | 251 | |||
AK195692.1 | - | 2, 3 | 91 | |||
BC017690.1 | - | 3, 4 | 284 | 4238947 | ||
BC021488.1 | - | 3, 4 | 275 | 5344864 | ||
BC024421.1 | - | 3, 4 | 269 | 4987584 |