Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173866.3 | NP_776291.1 | 4, 5 | 526 | ||
GenBank mRNA | AK033424.1 | - | 4, 5 | 527 | ||
AK075894.1 | - | 4, 5 | 545 | |||
AK076250.1 | - | 4, 5 | 544 | |||
AK082030.1 | - | 4, 5 | 569 | |||
BC034219.1 | - | 4, 5 | 558 | 5065322 |