Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173866.3 | NP_776291.1 | 10 | 1351 | ||
GenBank mRNA | AK031958.1 | - | 10 | 1543 | ||
AK033424.1 | - | 10 | 1352 | |||
AK076250.1 | - | 10 | 1369 | |||
AK082030.1 | - | 10 | 1394 | |||
AK163409.1 | - | 2 | 265 | |||
BC034219.1 | - | 10 | 1383 | 5065322 |