Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc88a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176841.4 | NP_789811.2 | 30 | 5428 | ||
GenBank mRNA | AB087827.1 | - | 30 | 5252 | ||
AK082771.1 | - | 2 | 472 | |||
AK129310.1 | - | 17 | 3231 | |||
BC037020.1 | - | 12 | 1997 |