Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175362.2 | NP_780571.2 | 16 | 2394 | ||
GenBank mRNA | AK037968.1 | - | 2 | 259 | ||
AK131134.1 | - | 16 | 2394 | |||
AY135367.1 | - | 16 | 2305 | |||
BC096592.1 | - | 16 | 2380 | 5318165 |