Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133985.2 | NP_598746.2 | 13, 14 | 1614 | ||
GenBank mRNA | AK031790.1 | - | 12, 13 | 1111 | ||
AK075837.1 | - | 12, 13 | 1122 | |||
AK129291.1 | - | 13, 14 | 1609 | |||
BC036148.1 | - | 4, 5 | 364 | |||
BC046453.1 | - | 14, 15 | 1638 | |||
BC060645.1 | - | 13, 14 | 1597 | 6852021 |