Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133985.2 | NP_598746.2 | 9 | 1209 | ||
GenBank mRNA | AK031790.1 | - | 8 | 706 | ||
AK075837.1 | - | 8 | 717 | |||
AK129291.1 | - | 9 | 1204 | |||
BC046453.1 | - | 10 | 1233 | |||
BC060645.1 | - | 9 | 1192 | 6852021 |