Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042652.1 | NP_001036117.1 | 6 | 1184 | ||
NM_133851.3 | NP_598612.1 | 7 | 1283 | |||
GenBank mRNA | AF305710.1 | - | 7 | 864 | ||
AK042697.1 | - | 765 | ||||
AK044819.1 | - | 7 | 856 | |||
AK076028.1 | - | 7 | 853 | |||
BC004787.1 | - | 7 | 833 | 3588697 | ||
BC100392.1 | - | 7 | 1283 | 30110191 |