Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042652.1 | NP_001036117.1 | 7 | 1338 | ||
NM_133851.3 | NP_598612.1 | 8 | 1437 | |||
GenBank mRNA | AF305710.1 | - | 8 | 1018 | ||
AK042697.1 | - | 919 | ||||
AK044819.1 | - | 8 | 1010 | |||
AK076028.1 | - | 8 | 1007 | |||
AK088389.1 | - | 8 | 1037 | |||
BC004787.1 | - | 8 | 987 | 3588697 | ||
BC009096.1 | - | 593 | ||||
BC100392.1 | - | 8 | 1437 | 30110191 |