Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042652.1 | NP_001036117.1 | 9 | 1511 | ||
NM_133851.3 | NP_598612.1 | 10 | 1610 | |||
GenBank mRNA | AF305710.1 | - | 10 | 1191 | ||
AK042697.1 | - | 1092 | ||||
AK044819.1 | - | 10 | 1183 | |||
AK076028.1 | - | 10 | 1180 | |||
AK088389.1 | - | 10 | 1210 | |||
AK155960.1 | - | 1 | 3125 | |||
BC004787.1 | - | 10 | 1160 | 3588697 | ||
BC009096.1 | - | 766 | ||||
BC100392.1 | - | 10 | 1610 | 30110191 |