Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178650.3 | NP_848765.2 | 2 | 80 | ||
GenBank mRNA | AK040487.1 | - | 2 | 81 | ||
AK075803.1 | - | 2 | 67 | |||
AK088119.1 | - | 145 | ||||
AK131177.1 | - | 1 | 61 | |||
AK146426.1 | - | 2 | 74 | |||
AK153836.1 | - | 2 | 78 | |||
AK154444.1 | - | 1 | 58 | |||
BC094901.1 | - | 1 | 55 | |||
BQ829173.1 | - | 2 | 316 |