Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 5 | 617 | ||
GenBank mRNA | AK076116.1 | - | 5 | 573 | ||
AK079615.1 | - | 5 | 579 | |||
AK088757.1 | - | 4 | 544 | |||
AK136821.1 | - | 5 | 611 | |||
AK146282.1 | - | 5 | 565 | |||
AK163705.1 | - | 5 | 596 | |||
AK168252.1 | - | 5 | 567 | |||
AK178553.1 | - | 3 | 238 | |||
AK183681.1 | - | 3 | 238 | |||
AK201030.1 | - | 3 | 242 | |||
AY523555.1 | - | 4 | 538 | |||
BC016261.1 | - | 4 | 535 | 4920560 | ||
BC021864.1 | - | 5 | 557 | 5123958 | ||
BC021949.1 | - | 4 | 368 |