Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 7 | 769 | ||
GenBank mRNA | AK076116.1 | - | 7 | 725 | ||
AK079615.1 | - | 7 | 731 | |||
AK088757.1 | - | 6 | 696 | |||
AK136821.1 | - | 7 | 763 | |||
AK146282.1 | - | 7 | 717 | |||
AK163705.1 | - | 7 | 748 | |||
AK168252.1 | - | 7 | 719 | |||
AY523555.1 | - | 6 | 690 | |||
BC016261.1 | - | 6 | 687 | 4920560 | ||
BC021864.1 | - | 7 | 709 | 5123958 | ||
BC021949.1 | - | 6 | 520 |