Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134141.4 | NP_598902.1 | 9 | 1033 | ||
GenBank mRNA | AK076116.1 | - | 9 | 989 | ||
AK079615.1 | - | 9 | 995 | |||
AK088757.1 | - | 8 | 960 | |||
AK136821.1 | - | 9 | 1027 | |||
AK146282.1 | - | 9 | 981 | |||
AK163705.1 | - | 9 | 1012 | |||
AK168252.1 | - | 9 | 983 | |||
AK182813.1 | - | 1 | 75 | |||
AY523555.1 | - | 8 | 954 | |||
BC016261.1 | - | 8 | 951 | 4920560 | ||
BC021864.1 | - | 9 | 973 | 5123958 | ||
BC021949.1 | - | 8 | 784 |