Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sephs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175400.6 | NP_780609.4 | 3, 4 | 670 | ||
GenBank mRNA | AK044971.1 | - | 3, 4 | 607 | ||
AK053638.1 | - | 3, 4 | 705 | |||
BC065165.1 | - | 3, 4 | 631 | 6417054 | ||
BC066037.1 | - | 3, 4 | 669 | 6405661 |