Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181406.3 | NP_852071.2 | 15, 16 | 1344 | ||
GenBank mRNA | AK075721.1 | - | 15, 16 | 1325 | ||
AK156182.1 | - | 15, 16 | 1322 | |||
AK166986.1 | - | 15, 16 | 1856 | |||
BC024878.1 | - | 15, 16 | 1324 | 3989666 |