Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178651.3 | NP_848766.2 | 8 | 822 | ||
GenBank mRNA | AF263460.1 | - | 6 | 376 | ||
AK028885.1 | - | 8 | 789 | |||
AK035541.1 | - | 8 | 809 | |||
AK038559.1 | - | 7 | 728 | |||
AK082853.1 | - | 8 | 824 | |||
AK143277.1 | - | 8 | 823 | |||
AK149549.1 | - | 1 | 973 | |||
AY682914.1 | - | 9 | 815 | |||
BC026565.1 | - | 4 | 240 | |||
BC027806.1 | - | 8 | 584 | |||
BC031705.1 | - | 5 | 336 | |||
BC055773.1 | - | 5 | 275 | |||
BC078440.1 | - | 6 | 399 | |||
BC141008.1 | - | 9 | 788 | 9055620 |