Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178651.3 | NP_848766.2 | 16 | 1637 | ||
GenBank mRNA | AF263460.1 | - | 14 | 1191 | ||
AK028885.1 | - | 16 | 1604 | |||
AK033990.1 | - | 662 | ||||
AK035541.1 | - | 16 | 1624 | |||
AK038559.1 | - | 15 | 1543 | |||
AK082853.1 | - | 16 | 1639 | |||
AK143277.1 | - | 16 | 1638 | |||
AK149549.1 | - | 8 | 4088 | |||
AY682914.1 | - | 17 | 1630 | |||
BC023833.1 | - | 8 | 741 | |||
BC024849.1 | - | 3 | 370 | |||
BC026565.1 | - | 12 | 1055 | |||
BC027806.1 | - | 16 | 1399 | |||
BC031705.1 | - | 13 | 1151 | |||
BC055773.1 | - | 13 | 1090 | |||
BC078440.1 | - | 14 | 1214 | |||
BC141008.1 | - | 17 | 1603 | 9055620 |