Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178651.3 | NP_848766.2 | 7 | 782 | ||
GenBank mRNA | AF263460.1 | - | 5 | 336 | ||
AK028885.1 | - | 7 | 749 | |||
AK035541.1 | - | 7 | 769 | |||
AK038559.1 | - | 6 | 688 | |||
AK082853.1 | - | 7 | 784 | |||
AK143277.1 | - | 7 | 783 | |||
AK184127.1 | - | 3 | 184 | |||
AY682914.1 | - | 8 | 775 | |||
BC026565.1 | - | 3 | 200 | |||
BC027806.1 | - | 7 | 544 | |||
BC031705.1 | - | 4 | 296 | |||
BC055773.1 | - | 4 | 235 | |||
BC078440.1 | - | 5 | 359 | |||
BC141008.1 | - | 8 | 748 | 9055620 |