Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163263.1 | NP_001156735.1 | 3 | 649 | ||
NM_182999.2 | NP_892044.1 | 4 | 496 | |||
GenBank mRNA | AK048862.1 | - | 3 | 608 | ||
AK051278.1 | - | 3 | 608 | |||
AK053783.1 | - | 4 | 484 | |||
AK082529.1 | - | 3 | 552 | |||
AK085436.1 | - | 3 | 627 | |||
AK087860.1 | - | 3 | 573 | |||
AK139883.1 | - | 4 | 474 | |||
AK220492.1 | - | 3 | 565 | |||
BC052482.1 | - | 4 | 435 | 6308280 | ||
CJ052397.1 | - | 4 | 477 |