Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134156.2 | NP_598917.1 | 12, 13 | 1667 | ||
GenBank mRNA | AK142591.1 | - | 3, 4 | 383 | ||
AK154281.1 | - | 12, 13 | 1660 | |||
BC054830.1 | - | 12, 13 | 1612 | 6308412 | ||
DQ288940.1 | - | 12, 13 | 1387 |